Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001302489.1 | NP_001289418.1 | 4 | 364 | ||
NM_001302490.1 | NP_001289419.1 | 3 | 296 | |||
NM_001302491.1 | NP_001289420.1 | 4 | 392 | |||
NM_001302492.1 | NP_001289421.1 | 4 | 392 | |||
NM_001610.3 | NP_001601.1 | 4 | 392 | |||
GenBank mRNA | AK298122.1 | - | 4 | 347 | ||
AK298439.1 | - | 4 | 367 | |||
AK299382.1 | - | 4 | 359 | |||
AK300569.1 | - | 3 | 320 | |||
AK300911.1 | - | 4 | 359 | |||
AK300931.1 | - | 4 | 350 | |||
AK301860.1 | - | 3 | 254 | |||
AK303894.1 | - | 4 | 359 | |||
AK308512.1 | - | 2 | 611 | |||
AK311431.1 | - | 4 | 385 | |||
BC003160.1 | - | 4 | 337 | 2957907 | ||
BC093010.1 | - | 4 | 350 | 30332133 | ||
DA382854.1 | - | 4 | 350 | |||
DC337294.1 | - | 4 | 451 | |||
X12548.1 | - | 4 | 346 |