Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Cpa5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144537.3 | NP_653120.1 | 2 | 594 | ||
GenBank mRNA | AF466283.1 | - | 2 | 595 | ||
AK015256.1 | - | 2 | 595 | |||
BC138114.1 | - | 1 | 112 | 8861133 | ||
BC145151.1 | - | 1 | 112 |