Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pomk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029037.4 | NP_083313.1 | 5 | 720 | ||
GenBank mRNA | AK015374.1 | - | 5 | 713 | ||
AK032677.1 | - | 5 | 721 | |||
AK036348.1 | - | 5 | 727 | |||
AK043620.1 | - | 5 | 721 | |||
AK138952.1 | - | 5 | 729 | |||
AK160873.1 | - | 5 | 726 | |||
AK170994.1 | - | 5 | 721 | |||
BC027296.1 | - | 4 | 700 | 4013355 |