Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Snx16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127191.2 | NP_001120663.1 | 3 | 739 | ||
NM_029068.4 | NP_083344.3 | 3 | 739 | |||
GenBank mRNA | AK015872.1 | - | 3 | 700 | ||
AK032055.1 | - | 3 | 696 | |||
AK166422.1 | - | 3 | 713 | |||
BC019424.1 | - | 2 | 504 | 5052234 | ||
BC070431.1 | - | 682 |