Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Snx16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127191.2 | NP_001120663.1 | 2 | 636 | ||
NM_029068.4 | NP_083344.3 | 2 | 636 | |||
GenBank mRNA | AK015872.1 | - | 2 | 597 | ||
AK032055.1 | - | 2 | 593 | |||
AK043975.1 | - | 2 | 606 | |||
AK166422.1 | - | 2 | 610 | |||
BC019424.1 | - | 1 | 401 | 5052234 |