Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NCF4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000631.4 | NP_000622.2 | 7 | 789 | ||
NM_013416.3 | NP_038202.2 | 7 | 789 | |||
XM_017028808.1 | XP_016884297.1 | 7 | 1153 | |||
GenBank mRNA | AB025219.1 | - | 7 | 641 | ||
AB025220.1 | - | 7 | 641 | |||
AK223324.1 | - | 7 | 760 | |||
AK290924.1 | - | 7 | 711 | |||
BC002798.1 | - | 7 | 789 | 3634177 | ||
BT007346.1 | - | 7 | 605 | |||
BX355636.2 | - | 7 | 765 | |||
CR456528.1 | - | 7 | 686 | |||
CR542078.1 | - | 7 | 605 | |||
X77094.1 | - | 7 | 735 |