Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001302490.1 | NP_001289419.1 | 1 | 157 | ||
NM_001302491.1 | NP_001289420.1 | 1 | 157 | |||
NM_001302492.1 | NP_001289421.1 | 1 | 157 | |||
NM_001610.3 | NP_001601.1 | 1 | 157 | |||
GenBank mRNA | AK298122.1 | - | 1 | 112 | ||
AK299382.1 | - | 1 | 124 | |||
AK300569.1 | - | 1 | 115 | |||
AK300911.1 | - | 1 | 124 | |||
AK300931.1 | - | 1 | 115 | |||
AK301860.1 | - | 1 | 115 | |||
AK303894.1 | - | 1 | 124 | |||
AK308512.1 | - | 66 | ||||
AK311431.1 | - | 1 | 150 | |||
BC003160.1 | - | 1 | 102 | 2957907 | ||
BC093010.1 | - | 1 | 115 | 30332133 | ||
DA382854.1 | - | 1 | 115 | |||
DC337294.1 | - | 1 | 216 | |||
X12548.1 | - | 1 | 111 |