Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pik3cb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029094.3 | NP_083370.2 | 24 | 3487 | ||
GenBank mRNA | AK003230.1 | - | 8 | 902 | ||
AK081350.1 | - | 24 | 3679 | |||
AK090116.1 | - | 24 | 3364 | |||
AK154106.1 | - | 24 | 3472 | |||
AK165230.1 | - | 947 | ||||
BC127073.1 | - | 23 | 3136 | 40110218 |