Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atp13a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164366.1 | NP_001157838.1 | 15, 16 | 1714 | ||
NM_029097.2 | NP_083373.2 | 15, 16 | 1714 | |||
GenBank mRNA | AK156104.1 | - | 15, 16 | 1693 | ||
AK171305.1 | - | 15, 16 | 1667 | |||
AK171554.1 | - | 14, 15 | 1967 | |||
AK212892.1 | - | 1, 2 | 110 | |||
BC023746.1 | - | 15, 16 | 1689 | 5353766 | ||
BC042661.1 | - | 15, 16 | 1659 | 4192094 |