Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rrp7a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029101.4 | NP_083377.3 | 2 | 184 | ||
GenBank mRNA | AK003698.1 | - | 2 | 135 | ||
AK032596.1 | - | 147 | ||||
AK036247.1 | - | 129 | ||||
AK088456.1 | - | 2 | 163 | |||
AK146821.1 | - | 2 | 147 | |||
AK153565.1 | - | 2 | 143 | |||
AK159553.1 | - | 2 | 129 | |||
AK168189.1 | - | 2 | 147 | |||
AK168850.1 | - | 2 | 137 | |||
BC012523.1 | - | 2 | 121 | 3602183 | ||
FJ687153.1 | - | 2 | 117 |