Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rrp7a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029101.4 | NP_083377.3 | 7 | 1607 | ||
GenBank mRNA | AK003698.1 | - | 7 | 1558 | ||
AK032596.1 | - | 1570 | ||||
AK036247.1 | - | 1552 | ||||
AK088456.1 | - | 7 | 1586 | |||
AK146821.1 | - | 7 | 1570 | |||
AK153565.1 | - | 7 | 1566 | |||
AK159553.1 | - | 7 | 1552 | |||
AK168189.1 | - | 7 | 1570 | |||
AK168850.1 | - | 7 | 1560 | |||
AK202035.1 | - | 1 | 70 | |||
BC012523.1 | - | 7 | 1544 | 3602183 |