Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc173 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077684.1 | NP_001071152.1 | 2 | 156 | ||
GenBank mRNA | AK015894.1 | - | 2 | 200 | ||
AK016309.1 | - | 2 | 199 | |||
BC099565.1 | - | 2 | 220 | |||
BC125570.1 | - | 2 | 156 | 40130085 | ||
BC132491.1 | - | 2 | 156 | 40130768 | ||
BC145015.1 | - | 2 | 156 | 9053563 |