Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NDUFA4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002489.3 | NP_002480.1 | 2 | 214 | ||
GenBank mRNA | AF201077.1 | - | 1 | 138 | ||
AK024837.1 | - | 2 | 163 | |||
AK130950.1 | - | 2 | 154 | |||
BC101794.1 | - | 2 | 138 | 8069300 | ||
BC101796.1 | - | 2 | 138 | 8069302 | ||
BC105295.1 | - | 2 | 185 | 6739636 | ||
BG623521.1 | - | 215 | ||||
CR407618.1 | - | 2 | 85 | |||
CR541716.1 | - | 2 | 85 | |||
U94586.1 | - | 2 | 175 |