Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Etnk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029250.2 | NP_083526.2 | 5 | 1019 | ||
GenBank mRNA | AK016135.1 | - | 5 | 1001 | ||
AK031560.1 | - | 996 | ||||
AK033268.1 | - | 5 | 996 | |||
AK044502.1 | - | 5 | 1020 | |||
AK049827.1 | - | 5 | 733 | |||
AK051039.1 | - | 1029 |