Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tex29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029326.1 | NP_083602.1 | 5, 6 | 586 | ||
GenBank mRNA | AK006092.1 | - | 5, 6 | 590 | ||
BC049746.1 | - | 5, 6 | 586 | 6772475 |