Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rilpl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021430.2 | NP_067405.1 | 6, 7 | 1484 | ||
GenBank mRNA | AB041584.1 | - | 6, 7 | 1114 | ||
AK013469.1 | - | 3, 4 | 377 | |||
BC029965.1 | - | 6, 7 | 1162 | |||
BC051945.1 | - | 6, 7 | 1484 | 5716540 | ||
BC051946.1 | - | 6, 7 | 1414 | 6403995 | ||
BC058384.1 | - | 6, 7 | 1377 | 5718399 |