Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Cul5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001161618.1 | NP_001155090.1 | 2 | 566 | ||
NM_027807.3 | NP_082083.2 | 2 | 566 | |||
GenBank mRNA | AK014894.1 | - | 2 | 159 | ||
AK018437.1 | - | 2 | 547 | |||
AK031955.1 | - | 2 | 569 | |||
AK046030.1 | - | 2 | 537 | |||
AK162082.1 | - | 260 | ||||
BC019661.1 | - | 2 | 524 | |||
BC037054.1 | - | 2 | 214 | |||
BC075710.1 | - | 2 | 495 | 6312416 | ||
CJ045576.1 | - | 2 | 569 |