Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vwa5b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029401.1 | NP_083677.1 | 2, 3 | 259 | ||
GenBank mRNA | AK016425.1 | - | 2, 3 | 280 | ||
BC057376.1 | - | 1, 2 | 131 | |||
BC132477.1 | - | 2, 3 | 259 | 40130754 | ||
BC137851.1 | - | 2, 3 | 259 | 8860870 | ||
BC144974.1 | - | 3, 4 | 314 |