Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pank1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001114339.2 | NP_001107811.1 | 4 | 1640 | ||
NM_023792.2 | NP_076281.1 | 4 | 940 | |||
GenBank mRNA | AF200357.1 | - | 4 | 940 | ||
AF347700.1 | - | 4 | 1638 | |||
AK050222.1 | - | 4 | 941 | |||
AK184272.1 | - | 2 | 130 | |||
BC023496.1 | - | 4 | 862 | 5054239 |