Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pank1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001114339.2 | NP_001107811.1 | 3 | 1387 | ||
NM_023792.2 | NP_076281.1 | 3 | 687 | |||
GenBank mRNA | AF200357.1 | - | 3 | 687 | ||
AF347700.1 | - | 3 | 1385 | |||
AK017345.1 | - | 3 | 1254 | |||
AK050222.1 | - | 3 | 688 | |||
BC023496.1 | - | 3 | 609 | 5054239 |