Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Klf17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029416.2 | NP_083692.2 | 2 | 557 | ||
GenBank mRNA | AF508984.1 | - | 2 | 523 | ||
AK018361.1 | - | 2 | 523 | |||
AK139827.1 | - | 2 | 524 | |||
BC064748.1 | - | 2 | 501 | 30476790 |