Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klhl24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029436.3 | NP_083712.4 | 7, 8 | 1852 | ||
GenBank mRNA | AK015239.1 | - | 7, 8 | 1838 | ||
AK044892.1 | - | 7, 8 | 1824 | |||
AK046446.1 | - | 5, 6 | 930 | |||
AK163616.1 | - | 5, 6 | 990 | |||
BC021407.1 | - | 5, 6 | 1406 |