Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Depdc1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001172092.1 | NP_001165563.1 | 8 | 1735 | ||
NM_029523.3 | NP_083799.2 | 8 | 1735 | |||
GenBank mRNA | AK018060.1 | - | 1 | 258 | ||
AK146458.1 | - | 8 | 1704 | |||
BC005799.1 | - | 1 | 58 | |||
BC023934.1 | - | 8 | 1728 | 5342167 |