Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Slc35d3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029529.3 | NP_083805.1 | 1 | 750 | ||
GenBank mRNA | AK042761.1 | - | 1 | 502 | ||
AK044973.1 | - | 1 | 487 | |||
AK047610.1 | - | 1 | 751 | |||
BC139194.1 | - | 1 | 580 | 8862216 |