Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lamp5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029530.2 | NP_083806.2 | 1, 2 | 446 | ||
GenBank mRNA | AK018222.1 | - | 1, 2 | 145 | ||
AK045946.1 | - | 1, 2 | 121 | |||
CX240608.1 | - | 1, 2 | 446 |