Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc38a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199217.1 | NP_001186146.1 | 4 | 356 | ||
NM_001199218.1 | NP_001186147.1 | 4 | 376 | |||
NM_023805.3 | NP_076294.2 | 4 | 524 | |||
GenBank mRNA | AF159856.1 | - | 4 | 326 | ||
AK002607.1 | - | 4 | 342 | |||
AK035155.1 | - | 544 | ||||
BC010678.1 | - | 4 | 356 | |||
BC021324.1 | - | 4 | 410 | |||
BC054846.1 | - | 4 | 499 | 6493334 | ||
BC055339.1 | - | 4 | 376 | 6415115 | ||
CJ183778.1 | - | 4 | 383 |