Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Erp44 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029572.2 | NP_083848.1 | 5 | 483 | ||
GenBank mRNA | AK003217.1 | - | 5 | 452 | ||
AK141377.1 | - | 5 | 441 | |||
AK151497.1 | - | 5 | 484 | |||
AK160113.1 | - | 5 | 466 | |||
AK172973.1 | - | 425 | ||||
AK216793.1 | - | 2 | 94 | |||
BC019558.1 | - | 5 | 390 | 4192664 |