Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NEK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199397.1 | NP_001186326.1 | 18 | 2112 | ||
NM_012224.2 | NP_036356.1 | 17 | 2190 | |||
XM_006714228.1 | XP_006714291.1 | 18 | 2114 | |||
XM_011532003.1 | XP_011530305.1 | 18 | 2114 | |||
XM_011532005.2 | XP_011530307.1 | 18 | 2114 | |||
XM_017008249.1 | XP_016863738.1 | 17 | 2016 | |||
XM_017008250.1 | XP_016863739.1 | 16 | 2134 | |||
XM_017008251.1 | XP_016863740.1 | 17 | 2016 | |||
XM_017008252.1 | XP_016863741.1 | 15 | 2036 | |||
XM_017008253.1 | XP_016863742.1 | 17 | 1962 | |||
XM_017008254.1 | XP_016863743.1 | 10 | 1072 | |||
GenBank mRNA | AB067488.1 | - | 17 | 2026 | ||
AK027580.1 | - | 14 | 1266 | |||
CR933642.1 | - | 18 | 2067 |