Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NEK3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146099.1 | NP_001139571.1 | 5 | 697 | ||
NM_002498.2 | NP_002489.1 | 5 | 697 | |||
NM_152720.2 | NP_689933.1 | 5 | 716 | |||
NR_027415.2 | 5 | 697 | ||||
GenBank mRNA | AB072828.1 | - | 4 | 366 | ||
AK131359.1 | - | 4 | 1615 | |||
AK290259.1 | - | 5 | 429 | |||
AL833951.1 | - | 5 | 556 | |||
BC019916.1 | - | 5 | 424 | 5000918 | ||
BQ648157.1 | - | 5 | 633 | |||
CV029372.1 | - | 4 | 321 | |||
Z25434.1 | - | 2 | 96 | |||
Z29067.1 | - | 3 | 180 |