Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dok5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163686.1 | NP_001157158.1 | 5 | 788 | ||
NM_029761.4 | NP_084037.3 | 5 | 788 | |||
GenBank mRNA | AF418208.1 | - | 5 | 650 | ||
AK012430.1 | - | 5 | 464 | |||
AK044148.1 | - | 5 | 656 |