Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gper1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029771.3 | NP_084047.2 | 3 | 1625 | ||
GenBank mRNA | AK018203.1 | - | 3 | 1529 | ||
AK030375.1 | - | 3 | 1626 | |||
BC138598.1 | - | 3 | 1480 | 8861618 | ||
BC138616.1 | - | 3 | 1480 | 8861636 |
Set Membership: |
![]() |