- Order Status
- Quick Order
-
Don't have an account ? Create Account
Gene Symbol: Psapl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175249.3 | NP_780458.2 | 1 | 303 | ||
GenBank mRNA | AK009408.1 | - | 1 | 292 | ||
AK028455.1 | - | 1 | 268 | |||
AK078699.1 | - | 1 | 268 | |||
BC119540.2 | - | 1 | 268 | |||
BC148306.2 | - | 1 | 268 | |||
BY770175.1 | - | 1 | 304 |