Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: St5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001326.1 | NP_001001326.1 | 18 | 3322 | ||
NM_029811.2 | NP_084087.2 | 17 | 2071 | |||
GenBank mRNA | AK008100.1 | - | 10 | 1145 | ||
BC030391.1 | - | 14 | 1797 | 4950637 | ||
BC032266.1 | - | 10 | 1217 | |||
BC051034.1 | - | 18 | 3503 | 5345299 |