Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ankrd55 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168403.1 | NP_001161875.1 | 8 | 1247 | ||
NM_001168404.1 | NP_001161876.1 | 8 | 784 | |||
NM_029898.3 | NP_084174.2 | 7 | 1141 | |||
GenBank mRNA | AK045480.1 | - | 7 | 775 | ||
AK081197.1 | - | 8 | 785 | |||
AK138273.1 | - | 7 | 796 | |||
AK140417.1 | - | 8 | 1111 | |||
BC152317.1 | - | 7 | 768 | 40088531 | ||
BC152536.1 | - | 7 | 768 | 40088530 |