Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trat1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198297.3 | NP_938039.2 | 6 | 418 | ||
GenBank mRNA | AJ297969.1 | - | 6 | 436 | ||
AK021155.1 | - | 1 | 27 | |||
AK030807.1 | - | 6 | 601 | |||
AK138735.1 | - | 484 | ||||
BC104350.1 | - | 5 | 381 | 40049557 | ||
BC104351.1 | - | 6 | 418 | 40049558 |