Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trat1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198297.3 | NP_938039.2 | 2, 3 | 152 | ||
GenBank mRNA | AJ297969.1 | - | 2, 3 | 170 | ||
AK030807.1 | - | 2, 3 | 335 | |||
AK138735.1 | - | 218 | ||||
BC104351.1 | - | 2, 3 | 152 | 40049558 |