Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Defb41 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_183124.3 | NP_898947.1 | 2 | 125 | ||
GenBank mRNA | AK020304.1 | - | 2 | 126 | ||
DQ012027.1 | - | 2 | 125 |