Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lcn9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029959.2 | NP_084235.1 | 5 | 450 | ||
GenBank mRNA | AF435738.1 | - | 5 | 450 | ||
AK020305.1 | - | 5 | 452 |