Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 6030458C11Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029998.3 | NP_084274.1 | 4 | 548 | ||
GenBank mRNA | AK035859.1 | - | 4 | 533 | ||
AK036721.1 | - | 4 | 438 | |||
AK038561.1 | - | 4 | 533 | |||
AK054133.1 | - | 4 | 522 | |||
AK081625.1 | - | 4 | 514 | |||
AK144748.1 | - | 4 | 501 | |||
AK152195.1 | - | 4 | 541 | |||
AK162187.1 | - | 4 | 562 | |||
AK163412.1 | - | 4 | 528 | |||
AK171821.1 | - | 3 | 376 | |||
BC119058.1 | - | 4 | 483 | 8733807 | ||
BC137759.1 | - | 4 | 483 | 8860778 |