Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ascc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198007.2 | NP_932124.2 | 4 | 961 | ||
GenBank mRNA | AK052745.1 | - | 4 | 925 | ||
AK144867.1 | - | 4 | 971 | |||
BC023451.1 | - | 1 | 356 |