Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ninl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207204.2 | NP_997087.2 | 23 | 4070 | ||
GenBank mRNA | AK129253.1 | - | 22 | 3649 | ||
BC067071.1 | - | 20 | 3380 | 30615484 | ||
CO427181.1 | - | 6 | 631 |
Gene Symbol: Alg13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK012910.1 | - | 705 |