Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ralgps2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159965.1 | NP_001153437.1 | 7 | 681 | ||
NM_001159966.1 | NP_001153438.1 | 7 | 579 | |||
NM_001159967.1 | NP_001153439.1 | 7 | 681 | |||
NM_001159968.1 | NP_001153440.1 | 6 | 576 | |||
NM_023884.4 | NP_076373.3 | 7 | 621 | |||
GenBank mRNA | AK018622.1 | - | 7 | 623 | ||
AK019543.1 | - | 6 | 579 | |||
AK029049.1 | - | 7 | 622 | |||
AK029993.1 | - | 8 | 759 | |||
AK033549.1 | - | 7 | 577 | |||
AK036803.1 | - | 6 | 553 | |||
AK161080.1 | - | 7 | 648 | |||
AK165002.1 | - | 7 | 573 | |||
AK169368.1 | - | 7 | 599 | |||
BC043132.1 | - | 7 | 586 | 6811441 | ||
BC052663.1 | - | 7 | 598 | 30051209 |