Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NHS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136024.3 | NP_001129496.1 | 9 | 4249 | ||
NM_001291867.1 | NP_001278796.1 | 9 | 4844 | |||
NM_001291868.1 | NP_001278797.1 | 8 | 4186 | |||
NM_198270.3 | NP_938011.1 | 8 | 4781 | |||
XM_011545528.2 | XP_011543830.1 | 6 | 4393 | |||
GenBank mRNA | AY436752.1 | - | 8 | 4781 | ||
AY456992.1 | - | 9 | 4249 | |||
AY456993.1 | - | 8 | 4904 | |||
BC136415.1 | - | 9 | 4570 | 9020403 | ||
BC171763.1 | - | 9 | 4570 | 9054416 | ||
CR749300.1 | - | 3 | 1619 | |||
CR936788.1 | - | 8 | 3955 | |||
GQ988776.1 | - | 9 | 4509 |