Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rnase6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030098.2 | NP_084374.2 | 2 | 464 | ||
GenBank mRNA | AK020595.1 | - | 2 | 408 | ||
AK038708.1 | - | 3 | 780 | |||
AY545655.1 | - | 2 | 442 | |||
BC094892.1 | - | 2 | 418 | 6432787 | ||
BC132632.1 | - | 2 | 397 | 40130909 | ||
BC132634.1 | - | 2 | 396 | 40130911 |