Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Htra3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042615.2 | NP_001036080.1 | 1 | 732 | ||
NM_030127.3 | NP_084403.2 | 1 | 732 | |||
GenBank mRNA | AK020653.1 | - | 1 | 416 | ||
AK035194.1 | - | 1 | 735 | |||
AY037300.1 | - | 1 | 517 | |||
AY156509.1 | - | 1 | 424 | |||
AY280664.1 | - | 1 | 424 | |||
BC138587.1 | - | 1 | 456 | 8861607 | ||
BC138588.1 | - | 1 | 456 | 8861608 |