Gene Symbol: Pde6h | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023898.4 | NP_076387.2 | 4 | 401 | ||
GenBank mRNA | AK020922.1 | - | 4 | 402 | ||
BC057290.1 | - | 373 | ||||
BC058192.1 | - | 3 | 2161 | |||
BC115671.1 | - | 4 | 379 | 40104107 | ||
BC115672.1 | - | 4 | 379 | 40104108 |