Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Uvrag | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178635.3 | NP_848750.3 | 12 | 1347 | ||
GenBank mRNA | AB020334.1 | - | 11 | 952 | ||
AK030814.1 | - | 12 | 1278 | |||
AK043084.1 | - | 12 | 1325 | |||
AK078085.1 | - | 12 | 1339 | |||
AK089900.1 | - | 12 | 1332 | |||
AK154903.1 | - | 12 | 1347 | |||
AK165015.1 | - | 12 | 1337 | |||
AK165185.1 | - | 12 | 1328 | |||
AK186606.1 | - | 3 | 208 | |||
AK202347.1 | - | 1 | 104 | |||
AK213196.1 | - | 3 | 208 | |||
BC034072.1 | - | 7 | 709 | |||
BC034176.1 | - | 12 | 1316 | 4954871 |