Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Csgalnact2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030165.3 | NP_084441.3 | 5 | 1518 | ||
GenBank mRNA | AK019496.1 | - | 5 | 1521 | ||
AK028045.1 | - | 5 | 1499 | |||
AK145992.1 | - | 5 | 1505 | |||
AK160867.1 | - | 5 | 1511 | |||
BC060169.1 | - | 3 | 3121 |