Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ltn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081068.1 | NP_001074537.1 | 7, 8 | 1115 | ||
GenBank mRNA | AK019706.2 | - | 7, 8 | 1046 | ||
AK036725.1 | - | 7, 8 | 1045 | |||
AK165062.1 | - | 7, 8 | 1036 | |||
AK173009.1 | - | 7, 8 | 986 | |||
AK206959.1 | - | 1, 2 | 122 | |||
BC116272.2 | - | 7, 8 | 1069 | |||
BC116273.2 | - | 7, 8 | 1072 |