Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ltn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081068.1 | NP_001074537.1 | 10 | 1949 | ||
GenBank mRNA | AK019706.2 | - | 10 | 1883 | ||
AK036725.1 | - | 10 | 1879 | |||
AK165062.1 | - | 10 | 1870 | |||
AK173009.1 | - | 10 | 1820 | |||
BC116272.2 | - | 10 | 1903 | |||
BC116273.2 | - | 10 | 1906 |